Anti-IgG Goat Polyclonal Antibody (Cy2®)

Lieferant: Jackson Immuno Research
005-000-121
JACK005-000-121EA 140 EUR
JACK005-000-121
Anti-IgG Goat Polyclonal Antibody (Cy2®)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR