Anti-IgG Horse Polyclonal Antibody

Lieferant: Jackson Immuno Research
007-000-003
JACK007-000-003EA 0 EUR
JACK007-000-003
Anti-IgG Horse Polyclonal Antibody
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Unknown
Host: Horse
Isotype: IgG
Reactivity: Horse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR