Anti-IgG Rabbit Polyclonal Antibody

Lieferant: Jackson Immuno Research
008-000-007
JACK008-000-007EA 0 EUR
JACK008-000-007
Anti-IgG Rabbit Polyclonal Antibody
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Unknown
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR