Anti-IgG Mouse Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Jackson Immuno Research
012-000-050
JACK012-000-050EA 324 EUR
JACK012-000-050
Anti-IgG Mouse Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR