Anti-IgG Donkey Polyclonal Antibody (Cy2®)

Lieferant: Jackson Immuno Research
015-000-003
JACK015-000-003EA 310 EUR
JACK015-000-003
Anti-IgG Donkey Polyclonal Antibody (Cy2®)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: Unknown
Host: Donkey
Isotype: IgG
Reactivity: Donkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR