Anti-IgY Goat Polyclonal Antibody (Peroxidase)

Lieferant: Jackson Immuno Research
015-000-050
JACK015-000-050EA 324 EUR
JACK015-000-050
Anti-IgY Goat Polyclonal Antibody (Peroxidase)
Antikörper
Type:
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR