Anti-IgG Goat Polyclonal Antibody

Lieferant: Jackson Immuno Research
017-000-001
JACK017-000-001EA 50.6 EUR
JACK017-000-001
Anti-IgG Goat Polyclonal Antibody
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR