Anti-IgG Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
109-005-098
JACK109-005-098EA 340 EUR
JACK109-005-098
Anti-IgG Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AMCA (7-amino-4-methylcoumarin-3-acetic acid)
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR