Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Jackson Immuno Research
109-065-098
JACK109-065-098EA 323 EUR
JACK109-065-098
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR