Anti-IgM Fc5µ Goat Polyclonal Antibody (Biotin-SP (long spacer))

Lieferant: Jackson Immuno Research
109-067-043
JACK109-067-043EA 450 EUR
JACK109-067-043
Anti-IgM Fc5µ Goat Polyclonal Antibody (Biotin-SP (long spacer))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR