Anti-IgM Fc5µ Goat Polyclonal Antibody (Cy3®)

Lieferant: Jackson Immuno Research
109-167-043
JACK109-167-043EA 576 EUR
JACK109-167-043
Anti-IgM Fc5µ Goat Polyclonal Antibody (Cy3®)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR