Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Jackson Immuno Research
109-546-008
JACK109-546-008EA 492 EUR
JACK109-546-008
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR