F(ab')2 Anti-IgA alpha Goat Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Jackson Immuno Research
109-605-011
JACK109-605-011EA 335 EUR
JACK109-605-011
F(ab')2 Anti-IgA alpha Goat Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR