Anti-IgG Goat Polyclonal Antibody (Horseradish Peroxidase (HRP))

Lieferant: Jackson Immuno Research
111-036-047
JACK111-036-047EA 371 EUR
JACK111-036-047
Anti-IgG Goat Polyclonal Antibody (Horseradish Peroxidase (HRP))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR