Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 594)

Lieferant: Jackson Immuno Research
112-065-175
JACK112-065-175EA 0 EUR
JACK112-065-175
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 594)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR