Anti-IgM µ Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
112-155-075
JACK112-155-075EA 0 EUR
JACK112-155-075
Anti-IgM µ Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR