F(ab) Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 594)

Lieferant: Jackson Immuno Research
112-587-003
JACK112-587-003EA 0 EUR
JACK112-587-003
F(ab) Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 594)
Antikörper
Type:
Antigen: IgG2c
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR