Anti-IgG2b Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Jackson Immuno Research
115-035-207
JACK115-035-207EA 525 EUR
JACK115-035-207
Anti-IgG2b Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type:
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR