Anti-IgG3 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Jackson Immuno Research
115-055-209
JACK115-055-209EA 509 EUR
JACK115-055-209
Anti-IgG3 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR