Anti-IgG Goat Antibody

Lieferant: Jackson Immuno Research
115-065-044
JACK115-065-044EA 340 EUR
JACK115-065-044
Anti-IgG Goat Antibody
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR