Anti-IgM µ Goat Polyclonal Antibody (Biotin-SP (long spacer))

Lieferant: Jackson Immuno Research
115-065-075
JACK115-065-075EA 412 EUR
JACK115-065-075
Anti-IgM µ Goat Polyclonal Antibody (Biotin-SP (long spacer))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR