Anti-IgG2b Goat Polyclonal Antibody (Cy5®)

Lieferant: Jackson Immuno Research
115-065-207
JACK115-065-207EA 489 EUR
JACK115-065-207
Anti-IgG2b Goat Polyclonal Antibody (Cy5®)
Antikörper
Type:
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR