Anti-IgG Goat Polyclonal Antibody (Cy2®)

Lieferant: Jackson Immuno Research
115-095-003
JACK115-095-003EA 217 EUR
JACK115-095-003
Anti-IgG Goat Polyclonal Antibody (Cy2®)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR