Anti-IgM µ Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
115-155-075
JACK115-155-075EA 377 EUR
JACK115-155-075
Anti-IgM µ Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type:
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 405
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR