Anti-IgG Goat Antibody (Cy3®)

Lieferant: Jackson Immuno Research
115-165-2070
JACK115-165-2070EA 544 EUR
JACK115-165-2070
Anti-IgG Goat Antibody (Cy3®)
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR