Anti-IgG2b Goat Polyclonal Antibody (Cy5®)

Lieferant: Jackson Immuno Research
115-175-207
JACK115-175-207EA 527 EUR
JACK115-175-207
Anti-IgG2b Goat Polyclonal Antibody (Cy5®)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR