Anti-IgM µ Goat Polyclonal Antibody (Cy2®)

Lieferant: Jackson Immuno Research
115-225-075
JACK115-225-075EA 415 EUR
JACK115-225-075
Anti-IgM µ Goat Polyclonal Antibody (Cy2®)
Antikörper
Type:
Antigen: IgG1+IgG2a+IgG2b+IgG3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Fc γ
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR