Anti-IgG2b Goat Polyclonal Antibody (Rhodamine Red-X)

Lieferant: Jackson Immuno Research
115-295-207
JACK115-295-207EA 446 EUR
JACK115-295-207
Anti-IgG2b Goat Polyclonal Antibody (Rhodamine Red-X)
Antikörper
Type:
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR