Anti-IgG1+IgG2a+IgG2b+IgG3 Anti-IgG1+IgG2a+IgG2b+IgG3 Goat Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Jackson Immuno Research
115-605-164
JACK115-605-164EA 552 EUR
JACK115-605-164
Anti-IgG1+IgG2a+IgG2b+IgG3 Anti-IgG1+IgG2a+IgG2b+IgG3 Goat Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Type:
Antigen: IgG1+IgG2a+IgG2b+IgG3
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: Digoxin
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR