Anti-IgG F(ab')2 Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Jackson Immuno Research
115-606-072
JACK115-606-072EA 538 EUR
JACK115-606-072
Anti-IgG F(ab')2 Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Type:
Antigen:
Clonality: Monoclonal
Clone:
Conjugation: Cy3®
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: Digoxin
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR