Anti-Digoxin Anti-Digoxin Mouse Monoclonal Antibody

Lieferant: Jackson Immuno Research
200-002-156
JACK200-002-156EA 314 EUR
JACK200-002-156
Anti-Digoxin Anti-Digoxin Mouse Monoclonal Antibody
Antikörper
Type:
Antigen: Digoxin
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor®647
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR