Anti-IgG Mouse Monoclonal Antibody (Peroxidase)

Lieferant: Jackson Immuno Research
200-062-156
JACK200-062-156EA 380 EUR
JACK200-062-156
Anti-IgG Mouse Monoclonal Antibody (Peroxidase)
Antikörper
Type:
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Peroxidase
Epitope: Heavy and Light Chains
Host: Mouse
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR