F(ab')2 Anti-IgM µ Rabbit Goat Polyclonal Antibody (Peroxidase)

Lieferant: Jackson Immuno Research
309-036-043
JACK309-036-043EA 429 EUR
JACK309-036-043
F(ab')2 Anti-IgM µ Rabbit Goat Polyclonal Antibody (Peroxidase)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: F(ab')2
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR