F(ab')2 Anti-IgM µ Rabbit Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
309-156-043
JACK309-156-043EA 0 EUR
JACK309-156-043
F(ab')2 Anti-IgM µ Rabbit Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Alkaline Phosphatase
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Chicken IgY (IgG) (H+L)
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR