Anti-IgG Rabbit Antibody (AMCA)

Lieferant: Jackson Immuno Research
315-155-045
JACK315-155-045EA 0 EUR
JACK315-155-045
Anti-IgG Rabbit Antibody (AMCA)
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: AMCA
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR