Anti-IgG Rabbit Antibody (Cy3®)

Lieferant: Jackson Immuno Research
315-165-003
JACK315-165-003EA 275 EUR
JACK315-165-003
Anti-IgG Rabbit Antibody (Cy3®)
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR