Anti-IgG Rat Antibody (Cy5®)

Lieferant: Jackson Immuno Research
415-175-166
JACK415-175-166EA 0 EUR
JACK415-175-166
Anti-IgG Rat Antibody (Cy5®)
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR