Anti-IgG Sheep Antibody

Lieferant: Jackson Immuno Research
515-005-071
JACK515-005-071EA 230 EUR
JACK515-005-071
Anti-IgG Sheep Antibody
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR