Anti-IgG Sheep Antibody

Lieferant: Jackson Immuno Research
515-225-062
JACK515-225-062EA 296 EUR
JACK515-225-062
Anti-IgG Sheep Antibody
Antikörper
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: Heavy and Light Chain
Host: Sheep
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR