Anti-IgY Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Jackson Immuno Research
703-056-155
JACK703-056-155EA 403 EUR
JACK703-056-155
Anti-IgY Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type:
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR