Anti-IgY Donkey Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
703-155-155
JACK703-155-155EA 269 EUR
JACK703-155-155
Anti-IgY Donkey Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin-SP
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR