Anti-IgG Donkey Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))

Lieferant: Jackson Immuno Research
711-475-152
JACK711-475-152EA 353 EUR
JACK711-475-152
Anti-IgG Donkey Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AMCA (7-amino-4-methylcoumarin-3-acetic acid)
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR