Anti-IgG Donkey Polyclonal Antibody (Rhodamine Red-X)

Lieferant: Jackson Immuno Research
711-585-152
JACK711-585-152EA 332 EUR
JACK711-585-152
Anti-IgG Donkey Polyclonal Antibody (Rhodamine Red-X)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine Red-X
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR