Anti-IgG Bovine Polyclonal Antibody (Peroxidase)

Lieferant: Jackson Immuno Research
713-065-003
JACK713-065-003EA 246 EUR
JACK713-065-003
Anti-IgG Bovine Polyclonal Antibody (Peroxidase)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: Heavy and Light Chains
Host: Bovine
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR