Anti-IgM Donkey Antibody

Lieferant: Jackson Immuno Research
715-175-020
JACK715-175-020EA 0 EUR
JACK715-175-020
Anti-IgM Donkey Antibody
Antikörper
Type:
Antigen: IgM
Clonality:
Clone:
Conjugation:
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR