Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Jackson Immuno Research
715-545-151
JACK715-545-151EA 496 EUR
JACK715-545-151
Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR