Closed cup flash tester
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
K16592
KOEHK16592EA 21640 EUR
KOEHK16592
Closed cup flash tester
Flammpunktprüfer
Rapid tester provides rapid determinations of flash point or sustained burning qualities by using a small sample.
  • One minute test result with a 2 ml sample
  • Convenient semi-automatic operations for flash/no flash tests
Closed cup model specifically used for routine flash point tests from –30 to +300 °C, in quality assurance and environmental compliance testing as well as actual flashpoint for paints, fragrances, hydrocarbons and other liquids.
Standards: ASTM ; IP 303, ISO 3679, ISO 3680, ISO 9038; DOT CFR 49-173.115; IATA.
Lieferumfang: Includes an automatic temperature control with a °C selector switch, a syringe, an electronic timer, an integral NIST traceable thermometer (0 to 300 °C), and an external fuel cylinder valve for connecting to a customer-supplied fuel cylinder or other fuel source.