TBX agar
Lieferant: LIOFILCHEM
10522
LIOF10522EA
34.3
EUR
LIOF10522
TBX agar
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
X-glucuronide, contained in the TBX agar formulation, is the chromogenic agent that allows the determination of the β-Dglucuronidase activity, which is a highly specific enzyme for E. coli. Gram-positive bacteria are inhibited by bile salts.