Media in bottles, sabouraud dextrose broth
Lieferant: LIOFILCHEM
402040
LIOF402040EA
45.2
EUR
LIOF402040
Media in bottles, sabouraud dextrose broth
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
Sabouraud dextrose broth (SDB) is a liquid medium used in qualitative procedures for isolation of yeasts and moulds and for the culture or subculture of fungi from clinical and nonclinical specimens.
Standards: This medium conforms to the requirements of the harmonised method in the United States Pharmacopoeia (USP), European Pharmacopoeia (Ph. Eur.) and Japanese Pharmacopoeia (JP) for the microbiological examination of non sterile products.