Orange Serum Agar
Lieferant: LIOFILCHEM
452690
LIOF452690EA
245
EUR
LIOF452690
Orange Serum Agar
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
Orange Serum Agar is a medium used for the isolation, cultivation and enumeration of microorganisms associated with spoilage of citrus fruits and derivates, such as lactic acid and acetic acid bacteria, yeasts and moulds.